Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1899MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1899MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1899MI Supplier MilliporeSigma Supplier No. AB1899

Rabbit Polyclonal Antibody

This Anti-Caspase 3 Antibody, large subunit and proform is validated for use in western blotting for the detection of Caspase 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal liquid in PBS, pH 7.2, containing 50% glycerol, 1% BSA and 0.02% thimerosal.
Gene Symbols CASP3; CPP-32; Yama; CASP-3; CPP32B; SCA-1; CPP32; EC 3.4.22.56 [Contains: Caspase-3 subunit p17 Caspase-3 subunit p12].
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004346.3
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.