Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3623MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3623MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3623MI Supplier MilliporeSigma Supplier No. AB3623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Caspase 3 Antibody, active (cleaved) form detects level of Caspase 3 and has been published and validated for use in Immunofluorescence (IF), Immunohistochemistry (immunohistochemistry), and Western Blot (WB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 3
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS, containing 50% glycerol, 1% BSA and 0.02% thimerosal.
Gene Symbols CASP3; CPP-32; Yama; CASP-3; CPP32B; SCA-1; CPP32; EC 3.4.22.56 [Contains: Caspase-3 subunit p17 Caspase-3 subunit p12].
Host Species Rabbit
Immunogen Cleavage specific synthetic peptide sequence (proprietary).
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004346.3
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.