Learn More
MilliporeSigma™ anti-Caspase-3, Cleaved, Polyclonal Available on GSA/VA Contract for Federal Government customers only.
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC67950UG
Catalog No. PC67950UG
Add to cart
Description
Description
Specifically detects Caspase-3, Cleaved Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Caspase-3, Cleaved
Unconjugated
purified
a synthetic peptide corresponding to amino acids surrounding the Asp175 cleavage site of caspase-3
50μg
≤ -70°C
Polyclonal
Human, Murine, Rat
Immuno Blotting, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
In PBS, 50% glycerol, 1% BSA, pH 7.2.
Rabbit
IgG
RUO
Primary
Recognizes the ∽17kDa cleaved fragment of caspase-3.