Learn More
Description
Specifications
Specifications
| Antigen | Caspase 3 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 3CSP03 |
| Conjugate | Unconjugated |
| Formulation | Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide. |
| Gene Accession No. | NM_004346.3; NM_032991.2 |
| Gene Symbols | CASP3, apopain, CASP-3, procaspase3, CPP32B, SCA-1, CPP-32, Apopain, CPP32, Yama, EC 3.4.22.56 [Contains: Caspase-3 subunit p17, Caspase-3 subunit p12]. |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human caspase-3 protein |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.