Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Caspase-3 (Pro), Rabbit Monoclonal,
Supplier: MilliporeSigma™ 04440
Description

Specifications
Caspase-3 (Pro) | |
Monoclonal | |
In 60% storage buffer (50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 0.01% sodium azide and 0.05% BSA) and 40% glycerol. | |
Rabbit | |
100 μL | |
CASP3, apopain, CASP-3, procaspase3, CPP32B, SCA-1, CPP-32, Apopain, CPP32, Yama | |
−20°C for 24 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
NM_004346.3, NM_032991.2 | |
KLH-conjugated synthetic peptide composed of residues at the N-terminus of human Pro-Caspase-3. | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction