Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Caspase 7 (Mch 3), Clone: 7CSP03, same as 13.1.97, Epredia™ LabVision

Catalog No. MS1129P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
200 μL
5 Slides
500 μL
7 mL
Concentration:
1mg/mL
200μg/mL
250μg/100μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1129P1 500 μL 200μg/mL
MS1129P0 100 μL 200μg/mL
MS1129P 1 mL 200μg/mL
MS1129PCL 100 μL 250μg/100μL
MS1129PABX 200 μL 1mg/mL
MS1129R7 7 mL
MS1129PCS 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1129P1 Supplier Epredia Supplier No. MS1129P1

Mouse Monoclonal Antibody

Caspase 7 functions as an effector caspase that cleaves downstream substrates responsible for producing the morphological and biochemical changes associated with apoptosis. The alpha form of caspase 7 is 303 amino acids and gets cleaved upon activatiuon to 20 and 12kDa subunits. A 253 aa beta form, presumably a result of alternative splicing, lacks the cysteine protease active site (QACRG) and may function as a dominant negative regulator of apoptosis.

Host Species: Mouse
Clone: 7CSP03; same as 13.1.97
Isotype: IgG1/κ
Cross Reactivity: Human and Mouse. Others-not tested.
Epitope: Not determined
Immunogen: Recombinant full length human Caspase 7 protein.
Molecular Weight: 35kDa (Proform) and 20kDa (Cleavage fragments)
Positive Control: Jurkat cells. Tonsil.
Cellular Localization: Cytoplasmic

Immunohistochemistry (Formalin/Paraffin), Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Caspase 7 (Mch 3) Ab-3
Classification Monoclonal
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant full length human Caspase 7 protein.
Purification Method Jurkat cells, tonsil
Quantity 500 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2–8 degrees
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.