Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 9 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB16969MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB16969MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB16969MI Supplier MilliporeSigma Supplier No. AB16969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Caspase 9 using this Anti-Caspase 9 Antibody, proform and lg. subunit validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoaffinity chromatography purified immunoglobulin. Liquid in PBS, containing 0.02% sodium azide.
Gene Symbols CASP9; CASP-9; ICE-LAP6; APAF3; MCH6; CASPASE-9c; OTTHUMP00000044594; APAF-3; EC 3.4.22.62 [Contains: Caspase-9 subunit p35; Caspase-9 subunit p10].
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 41-56 of human caspase-9 (Duan et al. 1996; Srinivasula et al. 1996; Nijhawan et al. 1997). Recognizes proform (46 kDa); sequence analysis predicts cross-reactivity with mouse caspase-9 (≥90%).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001229.2; NM_032996.1
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.