Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspr Mouse anti-Rat, Clone: K65/35, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN69
Description
Anti-Caspr Antibody, clone K65/35 is an antibody against Caspr for use in immunohistochemistry and western blotting.
Specifications
Caspr | |
Monoclonal | |
Unconjugated | |
NP_114450 | |
Mouse | |
Protein G | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
K65/35 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
CNTNAP, Caspr1, caspr1, P190, p190, NRXN4, CASPR | |
Recombinant protein corresponding to rat Caspr. | |
100 μg | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction