Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Catenin Mouse anti-Human, Clone: 11E4, MilliporeSigma™

Catalog No. MAB2083MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2083MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2083MI Supplier MilliporeSigma Supplier No. MAB2083
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect γ-Catenin using this Anti-γ-Catenin Antibody, clone 11E4 validated for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen γ-Catenin
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11E4
Conjugate Unconjugated
Formulation Liquid at 1mg/mL in 0.02Mphosphate buffer, pH 7.6, 0.25M NaCl, containing 0.1% sodium azide.
Gene Accession No. NM_002230.1; NM_021991.1
Gene Symbols JUP, PKGB, CTNNG, ARVD12, PDGB, gamma-catenin, DP3, DPIII, Desmoplakin-3
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.