Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Catenin Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB610253 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610253 50 μg
BDB610254 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610253 Supplier BD Biosciences Supplier No. 610253
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

γ-Catenin (plakoglobin) was identified as a compenent of desmosomes where it associates with desmoglein. γ-Catenin and β-Catenin are closely related proteins that have significant homology with the Drosophila armadillo protein. In addition to complexing with E-Cadherin, γ-Catenin and β-Catenin have been observed in association with the intracellular domain of N-Cadherin. It has been proposed that one molecule of α-Catenin and at least one molecule of β-Catenin and γ-Catenin simultaneously bind to a single cadherin molecule. A 19 amino acid sequence of desmoglein (Dsg1) was found to be critical for binding of γ-Catenin. This region has significant homology to the catenin-binding domain of classical cadherins, thus suggesting a common mechanism for γ-Catenin localization at both adherens junctions and desmosomes.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen γ-Catenin
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human γ-Catenin aa. 553-738
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.