Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Β-Catenin Mouse anti-Human, Mouse, Rat, Clone: 2H4A7, MilliporeSigma™

Catalog No. 05613MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-613-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-613-MI Supplier MilliporeSigma Supplier No. 05613
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect β-Catenin also known as beta catenin with Anti-β-Catenin Antibody, clone 2H4A7 (Mouse Monoclonal Antibody), that has been demonstrated to work in immunoprecipitation, western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Catenin
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H4A7
Conjugate Unconjugated
Formulation Protein G purified IgG in buffer containing 0.1M Tris-Glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide
Gene Accession No. NP_001895
Gene Symbols CTNNB1, CTNNB, FLJ25606, Beta-catenin, PRO2286, FLJ37923, DKFZp686D02253
Host Species Mouse
Immunogen fusion protein corresponding to residues 1-100 of human β-Catenin
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for up to 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.