Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

δ-Catenin Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB611537 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611537 150 μg
BDB611536 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611537 Supplier BD Biosciences Supplier No. 611537
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The catenins (α, β, γ, δ, p120[ctn]) are cytoplasmic proteins that are related to the Drosophila Armadillo protein. Catenins may have dual roles since they are components of cell-cell adherens junctions and can translocate to the nucleus after stimulation of the Wingless (Wnt-1 homolog) signaling pathway. α-Catenin has two subtypes: αE-Catenin, which is expressed ubiquitously, and αN-Catenin, which is expressed in the nervous system. β-Catenin binds to the cytoplasmic tail of E-Cadherin at adherens junctions and has been implicated in Wnt-1 signaling. γ-Catenin is associated with desmoglein in desmosomes and is closely related to β-Catenin. p120[ctn] is related to both β- and γ-catenin and is a substrate of tyrosine kinases localized at adherens junctions. δ-catenin was identified by its ability to bind the Alzheimer's disease-related protein, presenilin-1. It is most closely related to p120[ctn] and the desmosomal protein, p0071. It contains 10 Armadillo (Arm) repeats, as compared to the 13 Arm repeats found in β-Catenin. δ-Catenin is expressed at high levels in the developing nervous system, where it may be involved in neuronal progenitor cell migration and dendrite development.

Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse δ-Catenin aa. 85-194
Formula Weight [Chemical]: 160kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen δ-Catenin
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse δ-Catenin aa. 85-194
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.