Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Β-Catenin Rabbit anti-Human, Mouse, Rat, Horse, Ox, Polyclonal, MilliporeSigma™

Catalog No. ABE208MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE208MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE208MI Supplier MilliporeSigma Supplier No. ABE208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-β-Catenin Antibody is a Rabbit Polyclonal Antibody for detection of β-Catenin also known as catenin (cadherin-associated protein) beta 1 (88kD) and has been validated in western blot, immunocytochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Catenin
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols CTNNB1; CTNNB; Beta-catenin; PRO2286
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the consensus GSK3 phosphorylation site of human β-Catenin.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001895
Target Species Equine, Human, Mouse, Ox, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.