Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Β-Catenin Rabbit anti-Human, Rat, Mouse, Rabbit, Bovine, Polyclonal, MilliporeSigma™

Catalog No. 06734MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-734-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-734-MI Supplier MilliporeSigma Supplier No. 06734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-β-Catenin Antibody (Rabbit Polyclonal Antibody) validated in immunocytochemistry, immunoprecipitation, western blotting to detect β-Catenin also known as catenin (cadherin-associated protein) beta 1 (88kD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Catenin
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Gene Symbols CTNNB1; CTNNB; Beta-catenin; PRO2286; FLJ37923; FLJ25606; DKFZp686D02253
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (CGG-SYLDSGIHSGATTTAPSLSGK) corresponding to the consensus GSK3 phosphorylation site of human β -Catenin (amino acids 29-49).
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001091679
Target Species Bovine, Human, Mouse, Rabbit, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Prior to removing the cap centrifuge the vial and gently mix the solution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.