Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Cathepsin D Clone: BC011,

Catalog No. IM03100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IM03100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IM03100UG Supplier MilliporeSigma™ Supplier No. IM03100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Cathepsin D Clone: BC011 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

Cathepsin D is a normal lysosomal proteinase which is expressed in all cells. It is an aspartyl proteinase with a pH optimum in the range of 3-5 and contains two N-linked oligosaccharides. Cathepsin D is synthesized as an inactive 52kDa pro-enzyme. Activation involves the proteolytic removal of the 43 amino acid pro-fragment and internal cleavage to generate the two chain form made up of 34 and 14kDa subunits. Cathepsin D contains the mannose-6-phosphate lysosomal localization signal which targets the enzyme to the lysosomal compartment where it functions in the normal degradation of proteins. In certain tumor cells, cathepsin D is abnormally processed and is secreted in its 52kDa precursor form. Numerous clinical studies as well as in vitro evidence suggest that cathepsin D plays an important role in malignant transformation and may be a useful prognostic indicator for breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin D
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BC011
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.
Host Species Mouse
Immunogen mature, human cathepsin D
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽52kDa latent form, the ∽48kDa mature form, and the ∽34kDa subunit of cathepsin D in breast carcinoma tissue extracts.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.