Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Cathepsin S, Polyclonal

Catalog No. 219384100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
21-938-4100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-938-4100UL Supplier MilliporeSigma™ Supplier No. 219384100UL

Rabbit Polyclonal Antibody

Specifically detects Cathepsin S Clone: in Human samples, and it is validated for Immuno Blotting

Cathepsin S is a cysteine proteinase expressed in a variety of cells. The latent pro-cathepsin S (37kDa) is processed to the active single chain form (28kDa). Unlike cathepsin B and cathepsin L, cathepsin S exists only in single chain form. The pro peptide contains 99 amino acids. Mature Cathepsin S is normally localized in the lysosomes where it plays a major role in intracellular protein catabolism. High levels of cathepsin S expression have been demonstrated for many human tumors including cancers of the kidney, lung, colon, breast and ovary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin S
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen recombinant, human cathepsin S fused to maltose binding protein and expressed in E. coli
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽37kDa latent and the ∽28kDa active forms of cathepsin S in human tumors.
Target Species Human
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.