Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathespin D Mouse anti-Human, Unlabeled, Clone: 49, BD

Catalog No. BDB610801 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610801 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610801 Supplier BD Biosciences Supplier No. 610801
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cathepsin D, an enzyme that degrades proteins, was originally cloned during the search of estrogen responsive genes in MCF-7 cells. Cathepsin D is synthesized as the 43kDa preprocathepsin D that is cleaved to form a 46kDa glycosylated procathepsin D. Procathepsin is then processed into a 44kDa active Cathepsin D. The active and mature forms undergo a further cleavage that yields 28kDa and 15kDa (heavy and light chains, respectively) fragments in SDS-PAGE. The heavy and light chains of Cathepsin D are released into the extracellular medium. The maturation process of Cathepsin D occurs through the transit from the endoplasmic reticulum, Golgi apparatus, and to the lysosomes. Estrogens stimulate cell proliferation in a number of tumor cell lines and anti-estrogen therapy is often used in the treatment of breast cancer patients. Therefore, Cathepsin D, which is estrogen inducible, may have a role during the pathogenesis of breast tumors. Additionally, several other roles have been proposed for this enzyme, such as tissue remodeling, tumor invasion, and embryo implantation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathespin D
Applications Western Blot
Classification Monoclonal
Clone 49
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Cathespin D
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.