Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cav1.2 calcium channel Mouse anti-Mouse, Human, Rat, Rabbit, Clone: L57/46, MilliporeSigma™

Catalog No. MAB13170MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13170MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB13170MI Supplier MilliporeSigma Supplier No. MAB13170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cav1.2 calcium channel Antibody, clone L57/46 is validated for use in western blot, immunohistochemistry for the detection of Cav1.2 calcium channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav1.2 calcium channel
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone L57/46
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_001129994
Gene Symbols CACH2, CACN2, TS, CACNL1A1, CaV1.2, CCHL1A1, Cav1.2
Host Species Mouse
Immunogen Recombinant protein corresponding to rabbit Cav1.2 calcium channel at the cytoplasmic domain proximal to the C-terminus.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.