Learn More
Description
Specifications
Specifications
| Antigen | Cav2.2 |
| Applications | Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). |
| Gene Symbols | CACNA1B; BIII; CACH5; Cav2.2; CACNN; CACNL1A5 |
| Host Species | Rabbit |
| Immunogen | Purified peptide CNB1 from 851-867 of alpha 1B subunit of rat brain voltage-gated calcium channel (Accession Q02294; Westenbroek et al. 1992) containing N-terminal lysine and tyrosine, conjugated to KLH with gluteraldehyde (Harlow & Lane 1988). |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.