Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cav3.2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB982650UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB982650UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB982650UL Supplier MilliporeSigma Supplier No. AB982650UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cav3.2 Antibody is an antibody against Cav3.2 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav3.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 x
Gene Symbols CACNA1H: CAC1H
Host Species Rabbit
Immunogen Purified peptide from amino acids 581-595 of rat Cav3.2
Purification Method Affinity Purified
Quantity 50 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.