Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caveolin-1 Rabbit anti-Human, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3830
Description
This Anti-Caveolin-1 Antibody, phospho-specific (Tyr14) is validated for use in immunoprecipitation, western blotting for the detection of Caveolin-1.
Specifications
Caveolin-1 | |
Polyclonal | |
Protein A and peptide affinity purified immunoglobulin. Liquid in 10mM sodium HEPES, pH 7.5, 150mM NaCl, 100μg/mL BSA, with 50% glycerol. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Monkey, Mouse, Rat |
Immunoprecipitation, Western Blot | |
Unconjugated | |
CAV1; MSTP085; CAV; VIP21; Caveolin-1. | |
KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Tyr14 of human caveolin-1 | |
100 μL | |
NM_001753.3 | |
-20°C for 12 months, do not aliquot. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction