Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
calcium binding atopy-related autoantigen 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CBARA1.
Supplier: Abnova Corporation H00010367A01
Description
Sequence: DTALSFYHMAGASLDKVTMQQVARTVAKVELSDHVCDVVFALFDCDGNGELSNKEFVSIMKQRLMRGLEKPKDMGFTRLMQAMWKCAQETAWDFALPKQSpecifications
calcium binding atopy-related autoantigen 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CBARA1. | |
CBARA1 | |
CALC/DKFZp564C246/EFHA3/FLJ12684 | |
Mouse | |
50 μL | |
Yes | |
10367 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006077 | |
CBARA1 | |
CBARA1 (NP_006068, 380 a.a. ∼ 478 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human, Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction