Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbl Mouse anti-Human, Mouse, Rat, Clone: 7G10, MilliporeSigma™

Catalog No. 05440MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-440-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-440-MI Supplier MilliporeSigma Supplier No. 05440
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cbl Antibody, clone 7G10 is validated for use in immunoprecipitation, western blot, IC for the detection of Cbl.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cbl
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7G10
Conjugate Unconjugated
Formulation Purified IgG1k in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH7.4
Gene Accession No. NM_005188
Gene Symbols CBL, CBL2, RNF55, C-CBL
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 878-906 of human Cbl
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.