Learn More
Description
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q. [provided by RefSeq
Specifications
Specifications
| Antigen | CBR1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4G11-1C7 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant CBR1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CBR1 |
| Gene Accession No. | BC002511 |
| Gene Alias | CBR/SDR21C1/hCBR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.