Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS, Mouse, Clone: 3E1, Abnova™

Catalog No. 89016466 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-466 Supplier Abnova Corporation Supplier No. H00000875M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CBS.

The protein encoded by this gene acts as a homotetramer to catalyze the conversion of homocysteine to cystathionine, the first step in the transsulfuration pathway. The encoded protein is allosterically activated by adenosyl-methionine and uses pyridoxal phosphate as a cofactor. Defects in this gene can cause cystathionine beta-synthase deficiency (CBSD), which can lead to homocystinuria. [provided by RefSeq

Sequence: MPSETPQAEVGPTGCPHRSGPHSAKGSLEKGSPEDKEAKEPLWIRPDAPSRCTWQLGRPASESPHHHTAPAKSPKILPDILKKIGDTPMVRINKIGKKFG

Specifications

Antigen CBS
Applications ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, KnockDown, Western Blot
Classification Monoclonal
Clone 3E1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CBS.
Formulation PBS with no preservative; pH 7.4
Gene CBS
Gene Accession No. NM_000071
Gene Alias HIP4
Gene Symbols CBS
Host Species Mouse
Immunogen CBS (NP_000062, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 875
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.