Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX-4 Mouse anti-Human, Mouse, Clone: 10H10.2, MilliporeSigma™

Catalog No. MAB11012MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11012MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB11012MI Supplier MilliporeSigma Supplier No. MAB11012

Mouse Monoclonal Antibody

Use Anti-CBX-4 Antibody, clone 10H10.2 (Mouse Monoclonal Antibody) validated in western blot, Mplex to detect CBX-4 also known as chromobox homolog 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX-4
Applications Western Blot
Classification Monoclonal
Clone 10H10.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in PBS containing 0.05% sodium azide.
Gene Accession No. NM_007625
Gene Symbols PC2, NBP16, Pc2, hPC2, hPc2
Host Species Mouse
Immunogen The immunogen was GST-tagged fusion protein containing amino acids 77-551 of murine CBX-4.
Purification Method Purified from ascites by protein G affinity chromatography.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.