Learn More
chromobox homolog 5 (HP1 alpha homolog, Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CBX5 protein.
Supplier: Abnova Corporation H00023468D01P
Description
This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The encoded product is involved in the formation of functional kinetochore through interaction with essential kinetochore proteins. The gene has a pseudogene located on chromosome 3. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: MGKKTKRTADSSSSEDEEEYVVEKVLDRRVVKGQVEYLLKWKGFSEEHNTWEPEKNLDCPELISEFMKKYKKMKEGENNKPREKSESNKRKSNFSNSADDIKSKKKREQSNDIARGFERGLEPEKIIGATDSCGDLMFLMKWKDTDEADLVLAKEANVKCPQIVIAFYEERLTWHAYPEDAENKEKETAKSSpecifications
chromobox homolog 5 (HP1 alpha homolog, Drosophila) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CBX5 protein. | |
CBX5 | |
HP1/HP1A | |
Rabbit | |
Affinity Purified | |
RUO | |
23468 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_012117.1 | |
CBX5 | |
CBX5 (NP_036249.1, 1 a.a. ∼ 191 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.