Learn More
chibby homolog 1 (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CBY1 protein.
Supplier: Abnova Corporation H00025776D01P
Description
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MPFFGNTFSPKKTPPRKSASLSNLHSLDRSTREVELGLEYGSPTMNLAGQSLKFENGQWIAETGVSGGVDRREVQRLRRRNQQLEEENNLLRLKVDILLDMLSESTAESHLMEKELDELRISRKRKSpecifications
chibby homolog 1 (Drosophila) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CBY1 protein. | |
CBY1 | |
C22orf2/CBY/HS508I15A/PGEA1/PIGEA-14/PIGEA14/arb1 | |
Rabbit | |
Affinity Purified | |
RUO | |
25776 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001002880.1 | |
CBY1 | |
CBY1 (NP_001002880.1, 1 a.a. ∼ 126 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.