Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC141 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521169
Description
A suggested positive control is SK-N-SH cell lysate. PA5-21169 can be used with blocking peptide PEP-1283.
Coiled-coil domain-containing protein 141 (CCDC141), also known as CAMDI, is a recently identified disrupted in schizophrenia 1 (DISC1)-binding protein that also associates with myosin II. CCDC141 preferentially associates with phosphomyosin II and causes an accumulation of phosphomyosin II at the centrosome in a DISC1-dependent manner. Knockdown of CCDC141 expression by RNAi led to severely impaired radial migration with disoriented chromosomes, suggesting that CCDC141 is required for radial migration through DISC1 and myosin II-mediated centrosome positioning during neuronal development.
Specifications
| CCDC141 | |
| Polyclonal | |
| Unconjugated | |
| CCDC141 | |
| 2610301F02Rik; CAMDI; CCDC141; coiled-coil domain containing 141; coiled-coil domain-containing protein 141; Coiled-coil protein associated with myosin II and DISC1; ENSMUSG00000075261 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285025, 545428 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6ZP82 | |
| CCDC141 | |
| An 18 amino acid peptide near the amino terminus of human CCDC141. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction