Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

coiled-coil domain containing 44, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-612 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-612 Supplier Abnova Corporation Supplier No. H00051204B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CCDC44 protein.

TACO1 is a mitochondrial translational activator required for efficient translation of cytochrome c oxidase (COX) subunit I (MTCO1; MIM 516030) (Weraarpachai et al., 2009), a component of complex IV of the mitochondrial respiratory complex.[supplied by OMIM

Sequence: MSAWAAASLSRAAARCLLARGPGVRAAPPRDPRPSHPEPRGCGAAPGRTLHFTAAVPAGHNKWSKVRHIKGPKDVERSRIFSKLCLNIRLAVKEGGPNPEHNSNLANILEVCRSKHMPKSTIETALKMEKSKDTYLLYEGRGPGGSSLLIEALSNSSHKCQADIRHILNKNGGVMAVGARHSFDKKGVIVVEVEDREKKAVNLERALEMAIEAGAEDVKETEDEEERNVFKFICDASSLHQVRKKLDSLGLCSVSCALEFIPNSKVQLAEPDLEQAAHLIQALSNHEDVIHVYDNIE

Specifications

Antigen coiled-coil domain containing 44
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CCDC44 protein.
Formulation No additive
Gene CCDC44
Gene Accession No. NM_016360.2
Gene Symbols CCDC44
Host Species Mouse
Immunogen CCDC44 (NP_057444.2, 1 a.a. ∼ 297 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51204
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.