Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chemokine (C-C motif) ligand 14, Mouse, Clone: 3B12, Abnova™

Catalog No. 89000954 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-954 Supplier Abnova Corporation Supplier No. H00006358M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CCL14.

This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. Multiple transcript variants encoding different isoforms have been found for this gene. Read-through transcripts are also expressed that include exons from the upstream cytokine gene CCL15, and are represented as GeneID: 348249. [provided by RefSeq

Sequence: TKTESSSRGPYHPSECCFTYTTYKIPRQRIMDYYETNSQCSKPGIVFITKRGHSVCTNPSDKWVQDYIKDMKEN

Specifications

Antigen chemokine (C-C motif) ligand 14
Applications ELISA
Classification Monoclonal
Clone 3B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CCL14.
Formulation PBS with no preservative; pH 7.4
Gene CCL14
Gene Accession No. BC045165
Gene Alias CC-1/CC-3/CKb1/HCC-1/HCC-3/MCIF/NCC-2/NCC2/SCYA14/SCYL2/SY14
Gene Symbols CCL14
Host Species Mouse
Immunogen CCL14 (AAH45165, 20 a.a. ∼ 93 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6358
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.