Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chemokine (C-C motif) ligand 23, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-257 Supplier Abnova Corporation Supplier No. H00006368D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CCL23 protein.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist at CC chemokine receptor 1. Two alternatively spliced variants encoding different active isoforms have been identified. [provided by RefSeq

Sequence: MKVSVAALSCLMLVTALGSQARVTKDAETEFMMSKLPLENPVLLDMLWRRKIGPQMTLSHAAGFHATSADCCISYTPRSIPCSLLESYFETNSECSKPGVIFLTKKGRRFCANPSDKQVQVCVRMLKLDTRIKTRKN

Specifications

Antigen chemokine (C-C motif) ligand 23
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CCL23 protein.
Formulation PBS with no preservative; pH 7.4
Gene CCL23
Gene Accession No. NM_005064
Gene Alias CK-BETA-8/CKb8/Ckb-8/Ckb-8-1/MIP-3/MIP3/MPIF-1/SCYA23
Gene Symbols CCL23
Host Species Rabbit
Immunogen CCL23 (NP_005055.2, 1 a.a. ∼ 137 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 6368
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.