Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL23/MPIF-1 Mouse anti-Human, Clone: 69512, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB371
This item is not returnable.
View return policy
Description
CCL23/MPIF-1 Monoclonal antibody MAB371 Specifically detects CCL23/MPIF-1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization, Western BlottingSpecifications
CCL23/MPIF-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCL23 | |
E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120 Accession # P55773 | |
500 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Neutralization, Flow Cytometry, CyTOF | |
69512 | |
Western Blot 1 ug/mL, Neutralization 2-6 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P55773 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Detects human CCL23/MPIF-1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction