Learn More
chemokine (C-C motif) ligand 26, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CCL26 protein.
Supplier: Abnova Corporation H00010344D01P
Description
This gene is one of two Cys-Cys (CC) cytokine genes clustered on the q arm of chromosome 7. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for normal peripheral blood eosinophils and basophils. The product of this gene is one of three related chemokines that specifically activate chemokine receptor CCR3. This chemokine may contribute to the eosinophil accumulation in atopic diseases. [provided by RefSeq
Sequence: MMGLSLASAVLLASLLSLHLGTATRGSDISKTCCFQYSHKPLPWTWVRSYEFTSNSCSQRAVIFTTKRGKKVCTHPRKKWVQKYISLLKTPKQLSpecifications
chemokine (C-C motif) ligand 26 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CCL26 protein. | |
CCL26 | |
IMAC/MGC126714/MIP-4a/MIP-4alpha/SCYA26/TSC-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006072 | |
CCL26 | |
CCL26 (NP_006063.1, 1 a.a. ∼ 94 a.a) full-length human protein. | |
100 μg | |
Cytokines | |
10344 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.