Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
chemokine (C-C motif) ligand 4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CCL4 protein.
Supplier: Abnova Corporation H00006351D01P
Description
Sequence: MKLCVTVLSLLMLVAAFCSLALSAPMGSDPPTACCFSYTARKLPRNFVVDYYETSSLCSQPAVVFQTKRSKQVCADPSESWVQEYVYDLELNSpecifications
chemokine (C-C motif) ligand 4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CCL4 protein. | |
CCL4 | |
ACT2/AT744.1/G-26/LAG1/MGC104418/MGC126025/MGC126026/MIP-1-beta/MIP1B/MIP1B1/SCYA2/SCYA4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002984.2 | |
CCL4 | |
CCL4 (NP_002975.1, 1 a.a. ∼ 92 a.a) full-length human protein. | |
100 μg | |
Cell Adhesion | |
6351 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction