Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cyclin L1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CCNL1 protein.
Supplier: Abnova Corporation H00057018B01
Description
Sequence: MASGPHSTATAAAAASSAAPSAGGSSSGTTTTTTTTTGGILIGDRLYSEVSLTIDHSLIPEERLSPTPSMQDGLDLPSETDLRILGCELIQAAGILLRLPQVAMATGQVLFHRFFYSKSFVKHSFEIVAMACINLASKIEEAPRRIRDVINVFHHLRQLRGKRTPSPLILDQNYINTKNQVIKAERRVLKELGFCVHVKHPHKIIVMYLQVLECERNQTLVQTAWNYMNDSLRTNVFVRFQPETIACACIYLAARALQIPLPTRPHWFLLFGTTEEEIQEICIETLRLYTRKKPNYELLEKEVEKRKVALQEAKLKAKGLNPDGTPALSTLGGFSPASKPSSPREVKAEEKSPISINVKTVKKEPEDRQQASKSPYNGVRKDSKRSRNSRSASRSRSRTRSRSRSHTPRRHYNNRRSRSGTYSSRSRSRSRSHSESPRRHHNHGSPHLKAKHTRDDLKSSNRHGHKRKKSRSRSQSKSRDHSDAAKKHRHERGHHRDRRERSRSFERSHKSKHHGGSRSGHGRHRRSpecifications
cyclin L1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human CCNL1 protein. | |
CCNL1 | |
BM-001/PRO1073/ania-6a | |
Mouse | |
50 μL | |
Yes | |
57018 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_020307.2 | |
CCNL1 | |
CCNL1 (NP_064703.1, 1 a.a. ∼ 526 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction