Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR1 Mouse anti-Human, Alexa Fluor 647, Clone: 53504, BD

Catalog No. BDB557914 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557914 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557914 Supplier BD Biosciences Supplier No. 557914
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD191 (also known as CCR1, CC-CKR1, HM145 and MIP-1 α R), a seven-transmembrane, G-proteincoupled receptor, is the specific receptor for CC chemokine CCL3, CCL5, CCL7, CCL8, CCL14, CCL15, CCL16 and CCL23. It has been reported that human CD191 is expressed in peripheral monocytes and lymphocytes, bone marrow CD34+ cells. In addition, CD191 mRNA has been detected in dendritic cells and eosinophils. The human CD191 gene has been mapped to chromosome 3p21. The immunogen used to generate 53504 hybridoma was human CD191-transfected mouse cells. The antibody does not cross-react with human CCR5 transfected cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 53504
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description CCR1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.