Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR10 Mouse anti-Human, PerCP-Cy5.5, Clone: 1B5, BD

Catalog No. BDB564772 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564772 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564772 Supplier BD Biosciences Supplier No. 564772
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1B5 monoclonal antibody specifically binds to CCR10 (CC-chemokine receptor 10). CCR10 is a seven transmembrane domain G-protein-coupled receptor that is also known as GPR-2 (G-protein coupled receptor 2). CCR10 is expressed by cells that mediate epithelial immunity including a small subset of T memory cells, IgA-secreting B cells, dermal endothelial cells and fibroblasts. It is also expressed on EBV-immortalized B cells. Chemokine CCL27 and CCL28 are the ligands for CCR10. CCR10 reportedly not only functions in regulating the homing and function of cells that mediate epithelial immunity. It may also regulate the epithelial localization, survival, and proliferation of tumor cells and facilitate their evasion from immune surveillance.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR10
Applications Flow Cytometry
Classification Monoclonal
Clone 1B5
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description C-C CKR-10, C-C CKR-10, G-protein coupled receptor 2, CCR-10, GPR2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CCR10 Transfected Cell Line
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.