Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCR2 Polyclonal Antibody, DyLight™ 550
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523048
Description
CCR2 Polyclonal Antibody for ICC/IF, IHC (F), Flow
CCR2, a chemokine receptor, is a regulator of monocyte/macrophage traffickings and is upregulated in response to inflammation. Resistance to HIV-1 infection or delayed progression to AIDS may be linked to CCR2 polymorphisms. The ligand for CCR2 is monocyte chemoattractant protein (MCP-1), a chemokine produced by endothelial cells, smooth muscle cells, and macrophages. Activation of monocytes by MCP-1 may mediate the monocytic infiltration of tissues in atherosclerosis and other inflammatory disorders including rheumatoid arthritis, alveolitis, and tumor infiltration. Two isoforms are produced by alternative splicing. CCR2 has been reported to be expressed in a wide variety of tissues, including blood, brain, heart, kidney, liver, lung, ovary, pancreas, spinal cord, spleen, thymus, and vessel. Increased expression of CCR2 has been documented in CD3+ lymphocytes in rheumatoid arthritis. ESTs have been isolated from liver/spleen and prostate libraries.
Specifications
CCR2 | |
Polyclonal | |
DyLight 550 | |
Ccr2 | |
C-C chemokine receptor type 2; C-C CHEMOKINE RECEPTOR TYPE 2 (C-C CKR-2) (CC-CKR-2) (CCR-2) (CCR2) (JE/FIC RECEPTOR) (MCP-1 RECEPTOR); C-C CKR-2; C-C motif chemokine receptor 2; Cc-ckr2; CC-CKR-2; Ccr2; CCR-2; Ccr2a; CCR2B; CD192; CD192 antigen; chemoattractant protein-1 receptor; chemokine (C-C motif) receptor 2; chemokine (C-C) receptor 2; chemokine receptor CCR2; Ckr2; Ckr2a; Ckr2b; CMKBR2; hCG_14621; JE/FIC receptor; Mcp1 receptor; MCP-1 receptor; Mcp1r; MCP-1-R; MIP-1 alphaR; mJe-r; Monocyte chemoattractant protein 1 receptor; monocyte chemotactic protein 1 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12772, 729230 | |
4°C, store in dark | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
0.73 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
P41597, P51683 | |
Ccr2 | |
Synthetic peptide made to an N-terminal portion of the mouse CCR2 protein (within residues 20-100). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction