Learn More
anti-CCR3, PerCP, Clone: 83101, R&D Systems™
Description
Host Species: Rat
Clone: 83101
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: Y3 rat myeloid cell line transfected with mouse CCR3. Met1-Phe359
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | CCR3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 83101 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | P51678 |
| Gene Alias | b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 |
| Gene Symbols | CCR3 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.