Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD194 Mouse anti-Human, PE-Cy7, Clone: 1G1, BD

Catalog No. BDB561034 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561034 25 μg
BDB557864 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561034 Supplier BD Biosciences Supplier No. 561034
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 1G1 reacts with the human CC chemokine receptor, CCR4. CCR4 (previously known as K5-5), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CC chemokines, MDC/CCL22 and TARC/CC17. It has been shown that CCR4 mRNA is expressed mainly in thymus and spleen. CCR4 mRNA was also detected in peripheral T-lymphocytes, basophils, monocytes and platelets. In addition, CCR4 has been shown to be expressed selectively on polarized Th2 cells. The human CCR4 gene has been mapped to chromosome 3p24. The immunogen used to generate 1G1 hybridoma was human CCR4-transfected L1.2 mouse lymphoma cells. This antibody is NOT a neutralizing antibody.

Host Species: Mouse
Clone: 1G1
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human CCR4-Transfected L1.2 Mouse Lymphoma

Flow Cytometry, Immunofluorescence

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD194
Applications Flow Cytometry
Classification Monoclonal
Clone 1G1
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Description CCR4; C-C chemokine receptor type 4; CMKBR4; K5-5
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CCR4 Transfected Cell Line
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.