Learn More
CCR4 Mouse anti-Human, PE, Clone: 1G1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551120
Description
The monoclonal antibody 1G1 reacts with CD194, also known as the human CC Chemokine Receptor type 4 (CCR4). CCR4 is expressed on activated Th2 cells, regulatory T cells, activated NK cells, basophils, monocytes and platelets. CCR4 is a seven-transmembrane, G-protein-coupled receptor, and is the specific receptor for CC chemokines, CCL22/MDC/Macrophage-Derived Chemokine and CCL17/TARC/Thymus and Activation-Regulated Chemokine. It has been reported that CCR4 mRNA is expressed mainly in the thymus and spleen. The human CCR4 gene has been mapped to chromosome 3p24. The purified form of this antibody has been reported not to be a neutralizing antibody. The immunogen used to generate the 1G1 hybridoma has been reported to be human CCR4 transfected L1.2 mouse lymphoma cells.

Specifications
CCR4 | |
Monoclonal | |
0.2mg/mL | |
CCR4; C-C chemokine receptor type 4; CMKBR4; K5-5 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
1G1 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CCR4-Transfected L1.2 Mouse Lymphoma | |
0.2 mg | |
Immunology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.