Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR5, Fluorescein, Clone: CTC5, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Specifications
Antigen | CCR5 |
---|---|
Clone | CTC5 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1802F025
|
R&D Systems
FAB1802F025 |
25 Tests | N/A | N/A | N/A | ||||
FAB1802F100
|
R&D Systems
FAB1802F100 |
100 Tests | N/A | N/A | N/A | ||||
Description
CCR5 Monoclonal specifically detects CCR5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
CCR5 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human, Mouse, Rat | |
1234 | |
CHO Chinese hamster ovary cell line transfected with human CCR5 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
CTC5 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
CCR5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CCR5 in Western blots and human,mouse, and rat CCR5 in flow cytometry. For additional information regarding epitope specificity for this antibody and other R&D Systems anti-human CCR5 antibodies, see Reference 1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title