Learn More
CCR6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CCR6 peptide using ARM Technology.
Supplier: Abnova Corporation H00001235K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the beta chemokine receptor family, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. The gene is preferentially expressed by immature dendritic cells and memory T cells. The ligand of this receptor is macrophage inflammatory protein 3 alpha (MIP-3 alpha). This receptor has been shown to be important for B-lineage maturation and antigen-driven B-cell differentiation, and it may regulate the migration and recruitment of dentritic and T cells during inflammatory and immunological responses. Alternatively spliced transcript variants that encode the same protein have been described for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CCR6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CCR6 | |
CCR6 | |
A synthetic peptide of human CCR6 | |
RUO | |
1235 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
chemokine (C-C motif) receptor 6 | |
CCR6 | |
BN-1, CD196, CKR-L3, CKR6, CKRL3, CMKBR6, DCR2, DRY-6, GPR-CY4, GPR29, GPRCY4, STRL22 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.