Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR6, PE, Clone: 140706, R&D Systems™

Manufacturer:  R&D Systems FAB590P100

 View more versions of this product

Catalog No. FAB590P100


Add to cart

Description

Description

Host Species: Rat
Clone: 140706
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6. Met1-Met367

Applications:

Flow Cytometry

Specifications

Specifications

CCR6
140706
Flow Cytometry 10 uL/10^6 cells
O54689
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22
Rat
IgG2a
100 Tests
Primary
1235
Human, Mouse
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR6
RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6, Met1-Met367, Accession # O54689
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse CCR6-transfected rat RBL cells but not the parent cell line or cells transfected with mouse CCR5. Detects human CCR6 in flow cytometry.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit