Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR7 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
$162.80 - $444.00
Specifications
| Antigen | CCR7 |
|---|---|
| Clone | 4B12 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB3477A025
![]() |
R&D Systems
FAB3477A025 |
25 Tests |
Each for $162.80
|
|
|||||
FAB3477A100
![]() |
R&D Systems
FAB3477A100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
CCR7 Monoclonal specifically detects CCR7 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CCR7 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| CCR7 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse CCR7. |
| 4B12 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| NP_031745 | |
| 1236 | |
| RBC-2H3 cells expressing mouse CCR7 Accession # NP_031745 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title