Learn More
CCR9 Mouse anti-Human, Alexa Fluor 647, Clone: 112509, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 557975
Description
The 112509 monoclonal antibody specifically binds to the human CCR9 chemokine receptor. CCR9 is a thymus specific CC chemokine receptor expressed at high levels by essentially all CD4+ and CD8+ T lymphocytes. CCR9 and its ligand CCL25/TECK are involved in lymphocyte recruitment in the small intestine. The immunogen used to generate the 112509 hybridoma was human CCR9-transfected cells.

Specifications
CCR9 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CCR9 Transfected Cell Line | |
0.1 mg | |
Immunology | |
Human | |
IgG2a |
Flow Cytometry | |
112509 | |
Alexa Fluor 647 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.