Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR9, PerCP, Clone: 112509, R&D Systems™
Mouse Monoclonal Antibody
Specifications
Antigen | CCR9 |
---|---|
Clone | 112509 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB179C025
|
R&D Systems
FAB179C025 |
25 Tests | N/A | N/A | N/A | ||||
FAB179C100
|
R&D Systems
FAB179C100 |
100 Tests | N/A | N/A | N/A | ||||
Description
CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry.Specifications
CCR9 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
CCR9 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Specifically stains human CCR9 transfectants and MOLT-4 cells. It does not react with the parental BaF3 cells or human CCR7-transfected BaF3 cells. |
112509 | |
Flow Cytometry | |
PerCP | |
Mouse | |
P51686 | |
10803 | |
BaF3 mouse pro-B cell line transfected with human CCR9 Met1-Leu369 Accession # P51686 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title