Learn More
cell cycle related kinase, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase activates CDK2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
Specifications
| Antigen | cell cycle related kinase |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human CCRK protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CCRK |
| Gene Accession No. | BC002655.2 |
| Gene Alias | CDCH/p42 |
| Gene Symbols | CCRK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.