Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chemokine (C-C motif) receptor-like 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016961 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-961 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-961 Supplier Abnova Corporation Supplier No. H00009034B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CCRL2 protein.

This gene encodes a chemokine receptor like protein, which is predicted to be a seven transmembrane protein and most closely related to CCR1. Chemokines and their receptors mediated signal transduction are critical for the recruitment of effector immune cells to the site of inflammation. This gene is expressed at high levels in primary neutrophils and primary monocytes, and is further upregulated on neutrophil activation and during monocyte to macrophage differentiation. The function of this gene is unknown. This gene is mapped to the region where the chemokine receptor gene cluster is located. [provided by RefSeq

Sequence: MANYTLAPEDEYDVLIEGELESDEAEQCDKYDAQALSAQLVPSLCSAVFVIGVLDNLLVVLILVKYKGLKRVENIYLLNLAVSNLCFLLTLPFWAHAGGDPMCKILIGLYFVGLYSETFFNCLLTVQRYLVFLHKGNFFSARRRVPCGIITSVLAWVTAILATLPEFVVYKPQMEDQKYKCAFSRTPFLPADETFWKHFLTLKMNISVLVLPLFIFTFLYVQMRKTLRFREQRYSLFKLVFAIMVVFLLMWAPYNIAFFLSTFKEHFSLSDCKSSYNLDKSVHITKLIATTHCCINPLLYAFLDGTFSKYLCRCFHLRSNTPLQPRGQSAQGTSREEPDHSTEV

Specifications

Antigen chemokine (C-C motif) receptor-like 2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CCRL2 protein.
Formulation PBS with no preservative; pH 7.4
Gene CCRL2
Gene Accession No. NM_003965
Gene Alias CKRX/CRAM-A/CRAM-B/FLJ55815/HCR/MGC116710
Gene Symbols CCRL2
Host Species Mouse
Immunogen CCRL2 (NP_003956, 1 a.a. ∼ 344 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Membrane Receptors
Primary or Secondary Primary
Gene ID (Entrez) 9034
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.